Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dimethyl-2-thiohydantoin
1,3-Dimethyl-2-thiohydantoin 95%
1,3-Dimethyl-2-thiohydantoin 95%
1,3-Dimethyl-2-thiohydantoin 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Dimethyl-2-thiohydantoin 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1801-62-3
:
Formula:
C₅H₈N₂OS
MDL Number:
MFCD00186242
:
144.19 g/mol
Storage Temperature:
Ambient