Order Entry
Export
ContactUsLinkComponent
2,3-Dicyano-5-methylpyrazine 98%
2,3-Dicyano-5-methylpyrazine 98%
Catalog # APOSOR937582-5G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dicyano-5-methylpyrazine 98%
Catalog # APOSOR937582-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :144.1300 g/mol
Storage Temperature:Ambient
CAS Number:52197-12-3
Formula:C₇H₄N₄
MDL Number:MFCD00191635