Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S)-(-)-10-Mercaptoisoborneol
(1S)-(-)-10-Mercaptoisoborneol 95%
(1S)-(-)-10-Mercaptoisoborneol 95%
(1S)-(-)-10-Mercaptoisoborneol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
(1S)-(-)-10-Mercaptoisoborneol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
71242-58-5
:
Formula:
C₁₀H₁₈OS
MDL Number:
MFCD00151646
:
186.31 g/mol
Storage Temperature:
Ambient