Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Diisopropylphenol
2,4-Diisopropylphenol 95%
2,4-Diisopropylphenol 95%
2,4-Diisopropylphenol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Diisopropylphenol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2934-05-6
:
Formula:
C₁₂H₁₈O
MDL Number:
MFCD01707536
:
178.27
Storage Temperature:
Ambient