Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2,4-Dichlorophenyl)ethanol
2-(2,4-Dichlorophenyl)ethanol 98%
2-(2,4-Dichlorophenyl)ethanol 98%
2-(2,4-Dichlorophenyl)ethanol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(2,4-Dichlorophenyl)ethanol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
81156-68-5
:
Formula:
C₈H₈Cl₂O
MDL Number:
MFCD00192010
:
191.06 g/mol
Storage Temperature:
Ambient