0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,2-Dimethylbutyric acid
2,2-Dimethylbutyric acid 98%
2,2-Dimethylbutyric acid 98%
2,2-Dimethylbutyric acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Dimethylbutyric acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
595-37-9
:
Formula:
C₆H₁₂O₂
MDL Number:
MFCD00004200
:
116.16 g/mol
Storage Temperature:
Ambient