Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-Methyl 2,3-dichloropropionate
(±)-Methyl-2,3-dichloropropionate 95%
(±)-Methyl-2,3-dichloropropionate 95%
(±)-Methyl-2,3-dichloropropionate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
(±)-Methyl-2,3-dichloropropionate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
3674-09-7
:
Formula:
C₄H₆Cl₂O₂
MDL Number:
MFCD00000944
:
157 g/mol
Storage Temperature:
Ambient