Order Entry
Export
ContactUsLinkComponent
3-(1,3-Dioxolan-2-yl)benzonitrile 95%
3-(1,3-Dioxolan-2-yl)benzonitrile 95%
Catalog # APOSOR937002-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1,3-Dioxolan-2-yl)benzonitrile 95%
Catalog # APOSOR937002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.187 g/mol
Storage Temperature:Ambient
CAS Number:153329-04-5
Formula:C₁₀H₉NO₂
MDL Number:MFCD06208939