Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3,4-Dimethylphenyl)propionic acid
3-(3,4-Dimethylphenyl)propionic acid 95%
3-(3,4-Dimethylphenyl)propionic acid 95%
3-(3,4-Dimethylphenyl)propionic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(3,4-Dimethylphenyl)propionic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
25173-76-6
:
Formula:
C₁₁H₁₄O₂
MDL Number:
MFCD03425686
:
178.23 g/mol
Storage Temperature:
Ambient