0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Pyridine-2,3-dicarbonitrile
Pyridine-2,3-dicarbonitrile 95%
Pyridine-2,3-dicarbonitrile 95%
Pyridine-2,3-dicarbonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Pyridine-2,3-dicarbonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
17132-78-4
:
Formula:
C₇H₃N₃
MDL Number:
MFCD00191682
:
129.12 g/mol
Storage Temperature:
Ambient