Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Pentamethylene-1,3-dioxolan-4-one
2,2-Pentamethylene-1,3-dioxolan-4-one 95%
2,2-Pentamethylene-1,3-dioxolan-4-one 95%
2,2-Pentamethylene-1,3-dioxolan-4-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Pentamethylene-1,3-dioxolan-4-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
4423-79-4
:
Formula:
C₈H₁₂O₃
MDL Number:
MFCD00010852
:
156.18 g/mol
Storage Temperature:
Ambient