2,5-Dimethyl-3-hexyne-2,5-diol 95%
Catalog # APOSOR936618-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:142.2 g/mol
Storage Temperature:Ambient
CAS Number:142-30-3
Formula:C₈H₁₄O₂
MDL Number:MFCD00004468