Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethoxytoluene
3,5-Dimethoxytoluene 97%
3,5-Dimethoxytoluene 97%
3,5-Dimethoxytoluene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dimethoxytoluene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
4179-19-5
:
Formula:
C₉H₁₂O₂
MDL Number:
MFCD00015435
:
152.19 g/mol
Storage Temperature:
Ambient