Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-3-pyridineboronic acid
2,6-Dichloro-3-pyridineboronic acid 97%
2,6-Dichloro-3-pyridineboronic acid 97%
Irritant
2,6-Dichloro-3-pyridineboronic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dichloro-3-pyridineboronic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
148493-34-9
:
Formula:
C₅H₄BCl₂NO₂
MDL Number:
MFCD03094989
:
191.81 g/mol
Melting Pt:
97…99 °C
:
GHS07
Storage Temperature:
Ambient