1,2,3,4-Tetrahydropyrido[4,3-b][1,6]naphthyridine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:387358-40-9
:
Formula:C₁₁H₁₁N₃
MDL Number:MFCD00203189
:185.23 g/mol
Melting Pt:156…158 °C
:GHS07
Storage Temperature:Ambient