Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
Supplier:
Apollo Scientific
Certificates
Expand image
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
Supplier:
Apollo Scientific
About this item
CAS Number:
1363360-02-4
:
Formula:
C₁₀H₁₉NO
:
169.264 g/mol
Storage Temperature:
Ambient