
1-benzyl-1,4-diazepane 95+%
Catalog # APOSOR9337-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:190.29 g/mol
Boiling Pt:110 °C
Density:1.028 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4410-12-2
Formula:C₁₂H₁₈N₂
MDL Number:MFCD00800858