2,2'-Dipyrrolylmethane 95%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Di(1H-pyrrol-2-yl)methane
- CAS Number:21211-65-4
- Formula:C₉H₁₀N₂
- MDL Number:MFCD03931560
- Molecular Weight:146.19 g/mol
- Boiling Pt:120 °C/2.25mmHg
- Melting Pt:74 °C
- Storage Temperature:Refrigerator
4 Options Available Below
Product Details & Documents
Synonyms: Di(1H-pyrrol-2-yl)methane
- CAS Number:21211-65-4
- Formula:C₉H₁₀N₂
- MDL Number:MFCD03931560
- Molecular Weight:146.19 g/mol
- Boiling Pt:120 °C/2.25mmHg
- Melting Pt:74 °C
- Storage Temperature:Refrigerator



