Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-1,4-Cyclohexanediol
trans-1,4-Cyclohexanediol 97%
trans-1,4-Cyclohexanediol 97%
trans-1,4-Cyclohexanediol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
trans-1,4-Cyclohexanediol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
6995-79-5
:
Formula:
C₆H₁₂O₂
MDL Number:
MFCD00063612
:
116.16 g/mol
Storage Temperature:
Ambient