Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichloro-4-methoxypyrimidine
2,5-Dichloro-4-methoxypyrimidine 97%
2,5-Dichloro-4-methoxypyrimidine 97%
2,5-Dichloro-4-methoxypyrimidine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,5-Dichloro-4-methoxypyrimidine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
5750-74-3
:
Formula:
C₅H₄Cl₂N₂O
MDL Number:
MFCD11858598
:
179.01 g/mol
Storage Temperature:
Ambient