Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,O-Bis(trimethylsilyl)hydroxylamine
N,O-Bis(trimethylsilyl)hydroxylamine 95%
N,O-Bis(trimethylsilyl)hydroxylamine 95%
N,O-Bis(trimethylsilyl)hydroxylamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
N,O-Bis(trimethylsilyl)hydroxylamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
22737-37-7
:
Formula:
C₆H₁₉NOSi₂
MDL Number:
MFCD00008266
:
177.39 g/mol
Storage Temperature:
Ambient