Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibromo-5-iodobenzene
1,3-Dibromo-5-iodobenzene 95%
1,3-Dibromo-5-iodobenzene 95%
Irritant
1,3-Dibromo-5-iodobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,3-Dibromo-5-iodobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
19752-57-9
:
Formula:
C₆H₃Br₂I
MDL Number:
MFCD07778996
:
361.80 g/mol
Melting Pt:
120…130 °C
:
GHS07
Storage Temperature:
Ambient