Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octahydropyrrolo[3,4-c]pyrrole
Octahydropyrrolo[3,4-c]pyrrole 95%
Octahydropyrrolo[3,4-c]pyrrole 95%
Octahydropyrrolo[3,4-c]pyrrole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Octahydropyrrolo[3,4-c]pyrrole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
5840-00-6
:
Formula:
C₆H₁₂N₂
Storage Temperature:
Refrigerator