Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[3,2-b]pyridine-4-oxide
1H-Pyrrolo[3,2-b]pyridine-4-oxide 95%
1H-Pyrrolo[3,2-b]pyridine-4-oxide 95%
1H-Pyrrolo[3,2-b]pyridine-4-oxide 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1H-Pyrrolo[3,2-b]pyridine-4-oxide 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1116136-36-7
:
Formula:
C₇H₆N₂O
MDL Number:
MFCD12827548
:
134.14 g/mol
Storage Temperature:
Ambient