
2-(2,4-Dimethylphenoxy)acetohydrazide
Catalog # APOSOR9293-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:194.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:72293-69-7
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00156144