0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5,5-Dimethylpyrrolidin-2-one
5,5-Dimethylpyrrolidin-2-one 95%
5,5-Dimethylpyrrolidin-2-one 95%
5,5-Dimethylpyrrolidin-2-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5,5-Dimethylpyrrolidin-2-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
5165-28-6
:
Formula:
C₆H₁₁NO
MDL Number:
MFCD00128876
:
113.16 g/mol
Storage Temperature:
Ambient