
tert-Butyl-4,4-dimethyl-2,2-dioxooxathiazolidine-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:454248-55-6
:
Formula:C₉H₁₇NO₅S
MDL Number:MFCD22123261
:251.3 g/mol
Melting Pt:22 °C
:GHS07
Storage Temperature:Refrigerator