Order Entry
Export
ContactUsLinkComponent
2-Bromo-1,3-cyclohexanedione 95%
2-Bromo-1,3-cyclohexanedione 95%
Catalog # APOSOR928750-2G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-1,3-cyclohexanedione 95%
Catalog # APOSOR928750-2G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :191.03 g/mol
Storage Temperature:Ambient
CAS Number:60060-44-8
Formula:C₆H₇BrO₂
MDL Number:MFCD00971951