0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4,4-Dimethoxytetrahydro-4H-pyran
4,4-Dimethoxytetrahydro-4H-pyran 98%
4,4-Dimethoxytetrahydro-4H-pyran 98%
4,4-Dimethoxytetrahydro-4H-pyran 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4,4-Dimethoxytetrahydro-4H-pyran 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
28218-71-5
:
Formula:
C₇H₁₄O₃
MDL Number:
MFCD00040530
:
146.186 g/mol
Storage Temperature:
Ambient