Order Entry
Export
ContactUsLinkComponent
1,2-Dihydroxynaphthalene 95%
1,2-Dihydroxynaphthalene 95%
Catalog # APOSOR927003-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Dihydroxynaphthalene 95%
Catalog # APOSOR927003-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.17 g/mol
Storage Temperature:Ambient
CAS Number:574-00-5
Formula:C₁₀H₈O₂
MDL Number:MFCD00003959