Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichlorofuro[3,2-d]pyrimidine
2,4-Dichlorofuro[3,2-d]pyrimidine 95%
2,4-Dichlorofuro[3,2-d]pyrimidine 95%
:
Apollo Scientific
2,4-Dichlorofuro[3,2-d]pyrimidine 95%
:
Apollo Scientific
:
956034-07-4
:
:
C₆H₂Cl₂N₂O
:
MFCD11520866
:
188.9989 g/mol
:
GHS07
:
Ambient