Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5,6-dimethylpyrimidine
2,4-Dichloro-5,6-dimethylpyrimidine 98%
2,4-Dichloro-5,6-dimethylpyrimidine 98%
2,4-Dichloro-5,6-dimethylpyrimidine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichloro-5,6-dimethylpyrimidine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1780-32-1
:
Formula:
C₆H₆Cl₂N₂
MDL Number:
MFCD09910263
:
177.0300 g/mol
Storage Temperature:
Ambient