Trichloroacetyl isocyanate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3019-71-4
:
Formula:C₃Cl₃NO₂
MDL Number:MFCD00002033
:188.4 g/mol
Boiling Pt:80…85 °C
Density:1.59 g/cm³ (25 °C)
Flash Pt:65 °C
Storage Temperature:Refrigerator
UN:2206
ADR:6.1,III