Order Entry
Export
ContactUsLinkComponent
 
2,5-Dimethylterephthalonitrile 95%
Catalog # APOSOR925067-5G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethylterephthalonitrile 95%
Catalog # APOSOR925067-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :156.19 g/mol
Storage Temperature:Ambient
CAS Number:39095-25-5
Formula:C₁₀H₈N₂
MDL Number:MFCD00191411