Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethylterephthalic acid
2,5-Dimethylterephthalic acid 96%
2,5-Dimethylterephthalic acid 96%
2,5-Dimethylterephthalic acid 96%
Supplier:
Apollo Scientific
Certificates
Expand image
2,5-Dimethylterephthalic acid 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
6051-66-7
:
Formula:
C₁₀H₁₀O₄
MDL Number:
MFCD00020292
:
194.1840 g/mol
Storage Temperature:
Ambient