Acetanilide 95%
Bottle, 100 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N-Phenylacetamide
- CAS Number:103-84-4
- Formula:C₈H₉NO
- MDL Number:MFCD00008674
- Molecular Weight:135.17 g/mol
- Storage Temperature:Ambient
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:100 g



