Order Entry
Export
ContactUsLinkComponent
3,4-Dimethoxybenzoic acid 98%
3,4-Dimethoxybenzoic acid 98%
Catalog # APOSOR924272-500G
Supplier:  Apollo Scientific
CAS Number:  
3,4-Dimethoxybenzoic acid 98%
Catalog # APOSOR924272-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :182.18 g/mol
Storage Temperature:Ambient
CAS Number:93-07-2
Formula:C₉H₁₀O₄
MDL Number:MFCD00002500