Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(4-Chlorophenyl)morpholine
4-(4-Chlorophenyl)morpholine 95%
4-(4-Chlorophenyl)morpholine 95%
4-(4-Chlorophenyl)morpholine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-(4-Chlorophenyl)morpholine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
70291-67-7
:
Formula:
C₁₀H₁₂ClNO
MDL Number:
MFCD09038510
:
197.66 g/mol
Storage Temperature:
Ambient