Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methoxy-3,3-dimethyl-4-oxobutanoic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
32980-26-0
:
Formula:
C₇H₁₂O₄
MDL Number:
MFCD11617811
:
160.17 g/mol
Storage Temperature:
Ambient