2,3-Butanediol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:513-85-9
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00004523
:90.12 g/mol
Boiling Pt:180…184 °C
Melting Pt:19…25 °C
Density:1.002 g/cm³ (25 °C)
Flash Pt:85 °C
Storage Temperature:Ambient