4-(2-Hydroxyethoxy)benzaldehyde 95%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:22042-73-5
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00191450
- Molecular Weight:166.1739 g/mol
- Melting Pt:40 °C
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
- CAS Number:22042-73-5
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00191450
- Molecular Weight:166.1739 g/mol
- Melting Pt:40 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



