
7-Chloroimidazo[4,5-c]pyridine 95%
Catalog # APOSOR922610-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:5975-13-3
Formula:C₆H₄ClN₃
MDL Number:MFCD11501262