Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Diacetoxybenzene
1,2-Diacetoxybenzene 98%
1,2-Diacetoxybenzene 98%
1,2-Diacetoxybenzene 98%
APOSOR922181-25G
:
Apollo Scientific
:
1,2-Diacetoxybenzene 98%
APOSOR922181-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
194.19 g/mol
:
64 °C
:
GHS07
:
Ambient
:
635-67-6
:
C₁₀H₁₀O₄
:
MFCD00017219