2-Phenylethanol 95%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Boiling Pt:219…221 °C
- Melting Pt:–27 °C
- Density:1.02 g/cm³ (25 °C)
- Flash Pt:101–102 °C
- Storage Temperature:Ambient
- UN:2810
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Boiling Pt:219…221 °C
- Melting Pt:–27 °C
- Density:1.02 g/cm³ (25 °C)
- Flash Pt:101–102 °C
- Storage Temperature:Ambient
- UN:2810
- ADR:6.1,III



