Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Nonadien-1-ol (mixture of isomers)
3,6-Nonadien-1-ol (mixture of isomers) 95%
3,6-Nonadien-1-ol (mixture of isomers) 95%
3,6-Nonadien-1-ol (mixture of isomers) 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,6-Nonadien-1-ol (mixture of isomers) 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
76649-25-7
:
Formula:
C₉H₁₆O
MDL Number:
MFCD00036601
:
140.23 g/mol
Storage Temperature:
Ambient