1,5-Pentanediol 97%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane
- CAS Number:111-29-5
- Formula:C₅H₁₂O₂
- MDL Number:MFCD00002978
- Molecular Weight:104.15 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane
- CAS Number:111-29-5
- Formula:C₅H₁₂O₂
- MDL Number:MFCD00002978
- Molecular Weight:104.15 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



