Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethoxybenzene 97%

1,3-Diethoxybenzene 97%

Supplier:   Apollo Scientific
1,3-Diethoxybenzene 97%
Supplier:  Apollo Scientific
Certificates
1,3-Diethoxybenzene 97%

5 Options Available Below

Product Details & Documents
  • CAS Number:2049-73-2
  • Formula:C₁₀H₁₄O₂
  • MDL Number:MFCD00015145
  • Molecular Weight:166.22 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 49,20 €
    49,20 €
    1 * 25 g
       49,20 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 99,90 €
    99,90 €
    1 * 100 g
       99,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 474,00 €
    474,00 €
    1 * 500 g
       474,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 2 280,00 €
    2 280,00 €
    1 * 2,5 kg
       2 280,00 €

    Specifications:
    New Product
    New Product