Order Entry
Export
ContactUsLinkComponent
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
Catalog # APOSOR920336-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine 95%
Catalog # APOSOR920336-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.6000 g/mol
Storage Temperature:Ambient
CAS Number:50432-68-3
Formula:C₇H₆ClN₃
MDL Number:MFCD00222101