1,4-Benzoquinone dioxime 95%
Catalog # APOSOR920023-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:138.13 g/mol
Storage Temperature:Ambient
CAS Number:105-11-3
Formula:C₆H₆N₂O₂
MDL Number:MFCD00063636