Order Entry
Export
ContactUsLinkComponent
Methyl-4-acetoxybenzoate 95%
Methyl-4-acetoxybenzoate 95%
Catalog # APOSOR919737-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-acetoxybenzoate 95%
Catalog # APOSOR919737-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :194.19 g/mol
Storage Temperature:Ambient
CAS Number:24262-66-6
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00017517